Search
Search

ATZW10| Gene Symbol: ATZW10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128850.3 | NP_565757.2 | 3 - 4 | 1012 | 109 | |
| GenBank mRNA | AY069883.1 | - | 3 - 4 | 946 | 109 | |
| U80984.1 | - | 3 - 4 | 934 | 109 | ||
Set Membership: |
|