Search
Search

AT2G33000| Gene Symbol: AT2G33000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336421.1 | NP_001323452.1 | 1 - 2 | 588 | 131 | |
| NM_128860.3 | NP_180859.2 | 1 - 2 | 588 | 131 | ||
Set Membership: |
|