Search
Search

MSR2| Gene Symbol: MSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104041.5 | NP_175574.2 | 4 - 5 | 649 | 111 | |
| GenBank mRNA | AK229174.1 | - | 4 - 5 | 460 | 111 | |
| KY906048.1 | - | 4 - 5 | 411 | 111 | ||
Set Membership: |
|