Search
Search

SFR6| Gene Symbol: SFR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203747.1 | NP_001190676.1 | 2 - 3 | 694 | 115 | |
| NM_001340519.1 | NP_001329057.1 | 2 - 3 | 740 | 115 | ||
| NM_116730.5 | NP_192401.5 | 2 - 3 | 407 | 115 | ||
Set Membership: |
|