Search
Search

OEP80| Gene Symbol: OEP80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121967.4 | NP_568378.1 | 6 - 7 | 1270 | 96 | |
| GenBank mRNA | AF360205.1 | - | 6 - 7 | 1263 | 96 | |
| AY040053.1 | - | 6 - 7 | 1144 | 96 | ||
Set Membership: |
|