Search
Search

SNL5| Gene Symbol: SNL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198338.1 | NP_001185267.1 | 2 - 3 | 434 | 69 | |
| NM_001198339.2 | NP_001185268.1 | 2 - 3 | 620 | 69 | ||
| NM_001198340.2 | NP_001185269.1 | 2 - 3 | 603 | 69 | ||
| NM_104681.6 | NP_176197.3 | 2 - 3 | 620 | 69 | ||
Set Membership: |
|