Search
Search

SEC6| Gene Symbol: SEC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198445.1 | NP_001185374.1 | 25 - 26 | 2554 | 85 | |
| NM_105840.4 | NP_565026.1 | 25 - 26 | 2473 | 85 | ||
| GenBank mRNA | AY046014.1 | - | 25 - 26 | 2394 | 85 | |
| AY133819.1 | - | 24 - 25 | 2125 | 85 | ||
Set Membership: |
|