Search
Search

SUC1| Gene Symbol: SUC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105846.4 | NP_177333.1 | 2 - 3 | 1631 | 124 | |
| GenBank mRNA | AY049275.1 | - | 2 - 3 | 1481 | 124 | |
| BT020416.1 | - | 2 - 3 | 1327 | 124 | ||
| X75365.1 | - | 1386 | 124 | |||
Set Membership: |
|