Search
Search

ERO1| Gene Symbol: ERO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105887.4 | NP_177372.1 | 6 - 7 | 1083 | 75 | |
| GenBank mRNA | AK227807.1 | - | 6 - 7 | 912 | 75 | |
| BT008774.1 | - | 6 - 7 | 901 | 75 | ||
Set Membership: |
|