Search
Search

FIP1| Gene Symbol: FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335289.1 | NP_001318204.1 | 7 - 8 | 759 | 78 | |
| NM_001335290.1 | NP_001324184.1 | 6 - 7 | 579 | 78 | ||
| NM_001335291.1 | NP_001324181.1 | 7 - 8 | 628 | 78 | ||
| NM_001335292.1 | NP_001324182.1 | 7 - 8 | 624 | 78 | ||
| NM_001335293.1 | NP_001324183.1 | 7 - 8 | 611 | 78 | ||
| NM_126614.3 | NP_565332.1 | 7 - 8 | 633 | 78 | ||
| GenBank mRNA | AY086482.1 | - | 7 - 8 | 533 | 78 | |
Set Membership: |
|