Search
Search

ABH1| Gene Symbol: ABH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126934.4 | NP_565356.1 | 10 - 11 | 1154 | 68 | |
| GenBank mRNA | AF268377.1 | - | 10 - 11 | 1154 | 68 | |
| AF272891.1 | - | 10 - 11 | 1021 | 68 | ||
Set Membership: |
|