Search
Search

RECQI1| Gene Symbol: RECQI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337605.1 | NP_001327100.1 | 4 - 5 | 959 | 89 | |
| NM_111448.3 | NP_187225.2 | 5 - 6 | 1079 | 89 | ||
| GenBank mRNA | AJ404470.1 | - | 5 - 6 | 1046 | 89 | |
Set Membership: |
|