Search
Search

GSL10| Gene Symbol: GSL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337693.1 | NP_001326712.1 | 27 - 28 | 2891 | 154 | |
| NM_001337694.1 | NP_001326713.1 | 26 - 27 | 2759 | 154 | ||
| NM_111596.6 | NP_187372.5 | 26 - 27 | 2695 | 154 | ||
Set Membership: |
|