Search
Search

RABA5b| Gene Symbol: RABA5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111620.3 | NP_187397.1 | 1 - 2 | 401 | 66 | |
| GenBank mRNA | AY072444.1 | - | 1 - 2 | 401 | 66 | |
| BT002173.1 | - | 1 - 2 | 327 | 66 | ||
Set Membership: |
|