Search
Search

CIPK8| Gene Symbol: CIPK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118573.4 | NP_194171.1 | 12 - 13 | 1558 | 147 | |
| GenBank mRNA | AF290193.1 | - | 12 - 13 | 1158 | 147 | |
| BT000958.1 | - | 12 - 13 | 1158 | 147 | ||
Set Membership: |
|