Search
Search

ABCB28| Gene Symbol: ABCB28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036637.2 | NP_001031714.1 | 12 - 13 | 1718 | 63 | |
| NM_001084977.1 | NP_001078446.1 | 12 - 13 | 1711 | 63 | ||
| NM_118677.5 | NP_194275.2 | 12 - 13 | 1832 | 63 | ||
| GenBank mRNA | AK175486.1 | - | 12 - 13 | 1716 | 63 | |
| AY094470.1 | - | 12 - 13 | 1713 | 63 | ||
Set Membership: |
|