Search
Search

AT1G73875| Gene Symbol: AT1G73875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334610.1 | NP_001322223.1 | 6 - 7 | 814 | 84 | |
| NM_001334611.1 | NP_001322224.1 | 5 - 6 | 761 | 84 | ||
| NM_148650.4 | NP_683491.2 | 6 - 7 | 965 | 84 | ||
Set Membership: |
|