Search
Search

AT3G11960| Gene Symbol: AT3G11960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337966.1 | NP_001327782.1 | 4 - 5 | 617 | 66 | |
| NM_001337967.1 | NP_001327783.1 | 4 - 5 | 573 | 66 | ||
| NM_112029.4 | NP_187802.2 | 4 - 5 | 545 | 66 | ||
| GenBank mRNA | AK229623.1 | - | 4 - 5 | 545 | 66 | |
| BT006164.1 | - | 4 - 5 | 545 | 66 | ||
Set Membership: |
|