Search
Search

ARP5| Gene Symbol: ARP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202941.1 | NP_001189870.1 | 6 - 7 | 657 | 96 | |
| NM_112073.5 | NP_566422.4 | 6 - 7 | 702 | 96 | ||
| GenBank mRNA | FJ850974.1 | - | 6 - 7 | 552 | 96 | |
Set Membership: |
|