Search
Search

AGC1.5| Gene Symbol: AGC1.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112105.5 | NP_187875.1 | 3 - 4 | 1232 | 61 | |
| NM_202566.2 | NP_974295.1 | 2 - 3 | 1551 | 61 | ||
| NM_202567.2 | NP_974296.1 | 3 - 4 | 1311 | 61 | ||
| GenBank mRNA | AK229198.1 | - | 3 - 4 | 1177 | 61 | |
| BT011689.1 | - | 3 - 4 | 1164 | 61 | ||
| BT020332.1 | - | 1 - 2 | 867 | 61 | ||
Set Membership: |
|