Search
Search

REM1| Gene Symbol: REM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342099.1 | NP_001328023.1 | 4 - 5 | 1114 | 71 | |
| NM_119310.4 | NP_567880.1 | 4 - 5 | 1099 | 71 | ||
| GenBank mRNA | BT003891.1 | - | 4 - 5 | 1099 | 71 | |
| BT005002.1 | - | 4 - 5 | 898 | 71 | ||
Set Membership: |
|