Search
Search

AT4G32060| Gene Symbol: AT4G32060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085007.1 | NP_001078476.1 | 4 - 5 | 835 | 88 | |
| NM_119358.4 | NP_194934.1 | 4 - 5 | 987 | 88 | ||
| GenBank mRNA | AY072163.1 | - | 4 - 5 | 835 | 88 | |
| AY122982.1 | - | 4 - 5 | 779 | 88 | ||
Set Membership: |
|