Search
Search

AT4G32350| Gene Symbol: AT4G32350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342136.1 | NP_001328402.1 | 4 - 5 | 922 | 74 | |
| NM_001342137.1 | NP_001328401.1 | 6 - 7 | 644 | 74 | ||
| NM_119387.3 | NP_194961.2 | 5 - 6 | 836 | 74 | ||
| GenBank mRNA | AY080684.1 | - | 5 - 6 | 733 | 74 | |
| AY122956.1 | - | 5 - 6 | 563 | 74 | ||
Set Membership: |
|