Search
Search

APM1| Gene Symbol: APM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119463.5 | NP_195035.2 | 10 - 11 | 1558 | 80 | |
| GenBank mRNA | AK227105.1 | - | 10 - 11 | 1455 | 80 | |
| AY064961.1 | - | 10 - 11 | 1456 | 80 | ||
| AY143574.1 | - | 10 - 11 | 1431 | 80 | ||
| BT005811.1 | - | 10 - 11 | 1419 | 80 | ||
Set Membership: |
|