Search
Search

KUP5| Gene Symbol: KUP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119508.4 | NP_195079.2 | 2 - 3 | 782 | 62 | |
| GenBank mRNA | AY099556.1 | - | 2 - 3 | 612 | 62 | |
| BT010357.1 | - | 2 - 3 | 527 | 62 | ||
Set Membership: |
|