Search
Search

AT4G33770| Gene Symbol: AT4G33770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342232.1 | NP_001328707.1 | 1 - 2 | 714 | 101 | |
| NM_119535.5 | NP_195103.3 | 1 - 2 | 714 | 101 | ||
| GenBank mRNA | BT029235.1 | - | 1 - 2 | 314 | 101 | |
Set Membership: |
|