Search
Search

AT3G17100| Gene Symbol: AT3G17100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112586.3 | NP_566567.1 | 1 | 1046 | 114 | |
| NM_180270.3 | NP_850601.1 | 2 | 964 | 114 | ||
| GenBank mRNA | AK230300.1 | - | 1928 | 114 | ||
| AK317151.1 | - | 2 | 813 | 114 | ||
| AY045684.1 | - | 1 | 929 | 114 | ||
Set Membership: |
|