Search
Search

AT3G17340| Gene Symbol: AT3G17340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202987.1 | NP_001189916.1 | 20 - 21 | 3209 | 69 | |
| NM_112612.5 | NP_188360.3 | 20 - 21 | 3309 | 69 | ||
Set Membership: |
|