Search
Search

YSL5| Gene Symbol: YSL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338296.1 | NP_001325744.1 | 5 - 6 | 1173 | 92 | |
| NM_112646.2 | NP_566584.1 | 5 - 6 | 1208 | 92 | ||
| GenBank mRNA | AY515563.1 | - | 5 - 6 | 1089 | 92 | |
Set Membership: |
|