Search
Search

LPAT5| Gene Symbol: LPAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035647.3 | NP_001030724.2 | 2 - 3 | 826 | 92 | |
| NM_001084714.1 | NP_001078183.1 | 2 - 3 | 907 | 92 | ||
| NM_001084715.1 | NP_001078184.1 | 3 - 4 | 670 | 92 | ||
| NM_001338361.1 | NP_001326795.1 | 3 - 4 | 185 | 92 | ||
| NM_112771.5 | NP_188515.1 | 2 - 3 | 812 | 92 | ||
| NM_202606.2 | NP_974335.1 | 1 - 2 | 1021 | 92 | ||
| GenBank mRNA | AY099849.1 | - | 2 - 3 | 719 | 92 | |
| BT008462.1 | - | 1 - 2 | 551 | 92 | ||
Set Membership: |
|