Search
Search

SHM1| Gene Symbol: SHM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119954.4 | NP_195506.1 | 3 - 4 | 499 | 97 | |
| GenBank mRNA | AF428388.1 | - | 3 - 4 | 278 | 97 | |
| AJ271726.1 | - | 3 - 4 | 323 | 97 | ||
| AY057645.1 | - | 3 - 4 | 277 | 97 | ||
| AY070726.1 | - | 3 - 4 | 331 | 97 | ||
| BT006353.1 | - | 3 - 4 | 224 | 97 | ||
Set Membership: |
|