Search
Search

SHM1| Gene Symbol: SHM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119954.4 | NP_195506.1 | 5 - 6 | 698 | 66 | |
| GenBank mRNA | AF428388.1 | - | 5 - 6 | 477 | 66 | |
| AJ271726.1 | - | 5 - 6 | 522 | 66 | ||
| AY057645.1 | - | 5 - 6 | 476 | 66 | ||
| AY070726.1 | - | 5 - 6 | 530 | 66 | ||
| BT006353.1 | - | 5 - 6 | 423 | 66 | ||
Set Membership: |
|