Search
Search

AT4G38200| Gene Symbol: AT4G38200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342484.1 | NP_001328040.1 | 6 - 7 | 2691 | 88 | |
| NM_119981.4 | NP_195533.2 | 6 - 7 | 2713 | 88 | ||
Set Membership: |
|