Search
Search

AT1G02080| Gene Symbol: AT1G02080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197960.2 | NP_001184889.1 | 10 - 11 | 2062 | 133 | |
| NM_001331324.1 | NP_001321578.1 | 10 - 11 | 2048 | 133 | ||
| NM_100088.5 | NP_171710.4 | 11 - 12 | 2017 | 133 | ||
Set Membership: |
|