Search
Search

NAP1;3| Gene Symbol: NAP1;3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125077.4 | NP_568844.1 | 9 - 10 | 1008 | 98 | |
| GenBank mRNA | AF370487.1 | - | 9 - 10 | 976 | 98 | |
| AY064641.1 | - | 9 - 10 | 839 | 98 | ||
Set Membership: |
|