Search
Search

AT1G50140| Gene Symbol: AT1G50140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123988.1 | NP_001117460.1 | 22 - 23 | 2879 | 84 | |
| NM_001333421.1 | NP_001319186.1 | 24 - 25 | 3096 | 84 | ||
| NM_001333422.1 | NP_001319187.1 | 23 - 24 | 3023 | 84 | ||
| NM_001333423.1 | NP_001321914.1 | 24 - 25 | 2973 | 84 | ||
| NM_103899.4 | NP_175433.2 | 23 - 24 | 2945 | 84 | ||
Set Membership: |
|