Search
Search

HIT1| Gene Symbol: HIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198259.1 | NP_001185188.1 | 3 - 4 | 337 | 84 | |
| NM_001333440.1 | NP_001322424.1 | 2 - 3 | 567 | 84 | ||
| NM_001333441.1 | NP_001322422.1 | 3 - 4 | 366 | 84 | ||
| NM_001333442.1 | NP_001322423.1 | 3 - 4 | 322 | 84 | ||
| NM_103933.6 | NP_564573.3 | 3 - 4 | 463 | 84 | ||
| GenBank mRNA | AK228745.1 | - | 3 - 4 | 318 | 84 | |
Set Membership: |
|