Search
Search

IPT2| Gene Symbol: IPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128335.2 | NP_565658.1 | 8 - 9 | 1191 | 83 | |
| GenBank mRNA | AB062609.1 | - | 8 - 9 | 1103 | 83 | |
| AF109376.2 | - | 8 - 9 | 1165 | 83 | ||
| BT001906.1 | - | 8 - 9 | 1103 | 83 | ||
Set Membership: |
|