Search
Search

TIC| Gene Symbol: TIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125204.1 | NP_001118676.1 | 5 - 6 | 1719 | 110 | |
| NM_001338582.1 | NP_001326717.1 | 5 - 6 | 1719 | 110 | ||
| NM_113136.4 | NP_566705.1 | 5 - 6 | 1719 | 110 | ||
Set Membership: |
|