Search
Search

TOM20-1| Gene Symbol: TOM20-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203055.1 | NP_001189984.1 | 2 - 3 | 566 | 115 | |
| NM_113621.3 | NP_189343.1 | 3 - 4 | 450 | 115 | ||
| GenBank mRNA | AJ296023.1 | - | 3 - 4 | 397 | 115 | |
Set Membership: |
|