Search
Search

AT5G63640| Gene Symbol: AT5G63640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345600.1 | NP_001331835.1 | 4 - 5 | 1212 | 116 | |
| NM_125759.6 | NP_201169.1 | 4 - 5 | 634 | 116 | ||
| GenBank mRNA | AY062583.1 | - | 4 - 5 | 417 | 116 | |
| AY114647.1 | - | 2 - 3 | 184 | 116 | ||
Set Membership: |
|