Search
Search

PSAN| Gene Symbol: PSAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037056.2 | NP_001032133.2 | 1 - 2 | 500 | 90 | |
| NM_125800.5 | NP_201209.1 | 1 - 2 | 593 | 90 | ||
| GenBank mRNA | AY062835.1 | - | 1 - 2 | 350 | 90 | |
| AY081594.1 | - | 1 - 2 | 294 | 90 | ||
| U32176.1 | - | 1 - 2 | 324 | 90 | ||
Set Membership: |
|