Search
Search

AIL7| Gene Symbol: AIL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345694.1 | NP_001318882.1 | 3 - 4 | 853 | 87 | |
| NM_001345695.1 | NP_001330267.1 | 1 - 2 | 644 | 87 | ||
| NM_001345696.1 | NP_001330268.1 | 1 - 2 | 644 | 87 | ||
| NM_125949.7 | NP_201354.5 | 2 - 3 | 710 | 87 | ||
| GenBank mRNA | AY560887.1 | - | 1 - 2 | 536 | 87 | |
Set Membership: |
|