Search
Search

ACR1| Gene Symbol: ACR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345724.1 | NP_001330475.1 | 3 - 4 | 497 | 107 | |
| NM_125986.3 | NP_201390.1 | 3 - 4 | 358 | 107 | ||
| GenBank mRNA | AF360256.1 | - | 3 - 4 | 323 | 107 | |
| AF528058.1 | - | 2 - 3 | 170 | 107 | ||
| AY040070.1 | - | 2 - 3 | 119 | 107 | ||
Set Membership: |
|