Search
Search

bZIP58| Gene Symbol: bZIP58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101230.4 | NP_172817.2 | 1 | 637 | 78 | |
| GenBank mRNA | AB493455.1 | - | 1 | 465 | 78 | |
| AF332430.1 | - | 1 | 465 | 78 | ||
Set Membership: |
|