Search
Search

ATGA2OX1| Gene Symbol: ATGA2OX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106491.2 | NP_177965.1 | 2 - 3 | 837 | 65 | |
| GenBank mRNA | AJ132435.1 | - | 2 - 3 | 767 | 65 | |
| BT002763.1 | - | 2 - 3 | 727 | 65 | ||
Set Membership: |
|