Search
Search

SAR3| Gene Symbol: SAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106716.4 | NP_178183.2 | 2 - 3 | 2901 | 78 | |
| GenBank mRNA | AF411839.1 | - | 2 - 3 | 2775 | 78 | |
| AY942798.1 | - | 2 - 3 | 2564 | 78 | ||
Set Membership: |
|