Search
Search

PHT4;3| Gene Symbol: PHT4;3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035747.2 | NP_001030824.1 | 1 - 2 | 523 | 93 | |
| NM_001339292.1 | NP_001326351.1 | 1 - 2 | 627 | 93 | ||
| NM_114565.3 | NP_190282.2 | 1 - 2 | 531 | 93 | ||
| GenBank mRNA | BT015413.1 | - | 1 - 2 | 458 | 93 | |
Set Membership: |
|