Search
Search

DD46| Gene Symbol: DD46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102051.3 | NP_564154.1 | 2 - 3 | 291 | 85 | |
| GenBank mRNA | AK226416.1 | - | 2 - 3 | 292 | 85 | |
| KJ138995.1 | - | 2 - 3 | 197 | 85 | ||
Set Membership: |
|