Search
Search

AT5G22320| Gene Symbol: AT5G22320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203434.1 | NP_001190363.1 | 2 - 3 | 265 | 118 | |
| NM_122137.4 | NP_568416.1 | 2 - 3 | 358 | 118 | ||
| GenBank mRNA | AY056427.1 | - | 2 - 3 | 196 | 118 | |
| AY090323.1 | - | 2 - 3 | 171 | 118 | ||
Set Membership: |
|